missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorophenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 3900-89-8
Molecular Formula C6H6BClO2
Molecular Weight (g/mol) 156.38
MDL Number MFCD00674012
InChI Key RRCMGJCFMJBHQC-UHFFFAOYSA-N
Synonym 2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl
PubChem CID 2734322
IUPAC Name (2-chlorophenyl)boronic acid
SMILES B(C1=CC=CC=C1Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC358990010
SDS View Documents
Thermo Scientific Chemicals
358990010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC358990050
SDS View Documents
Thermo Scientific Chemicals
358990050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3900-89-8
156.38
RRCMGJCFMJBHQC-UHFFFAOYSA-N
2734322
B(C1=CC=CC=C1Cl)(O)O
C6H6BClO2
MFCD00674012
2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl
(2-chlorophenyl)boronic acid

Specifications

3900-89-8
100.0
96% min. (HPLC)
C6H6BClO2
1g
Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol
B(C1=CC=CC=C1Cl)(O)O
156.38
156.38
2-Chlorophenylboronic acid
96.0
Authentic
Glass bottle
MFCD00674012
2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl
RRCMGJCFMJBHQC-UHFFFAOYSA-N
(2-chlorophenyl)boronic acid
2734322
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only