missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 149140250

 View more versions of this product

Catalog No. AC149140250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Chlorophenylhydrazine hydrochloride
41052-75-9
100.0
96% min. (Argentometry)
C6H8Cl2N2
MFCD00012928
15, 422
Solubility in water: soluble
[H+].[Cl-].NNC1=CC=CC=C1Cl
179.04
CHEBI:1046
97%
97%
96.0
Authentic
Glass bottle
ClC6H4NHNH2·HCl
25g
2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride
ADODRSVGNHNKAT-UHFFFAOYSA-N
(2-chlorophenyl)hydrazine;hydrochloride
443287
179.05

Chemical Identifiers

41052-75-9
179.04
ADODRSVGNHNKAT-UHFFFAOYSA-N
443287
(2-chlorophenyl)hydrazine;hydrochloride
C6H8Cl2N2
MFCD00012928
2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride
CHEBI:1046
[H+].[Cl-].NNC1=CC=CC=C1Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Do not eat,drink or smoke when using this product.
IF

GHS Signal Word: Warning

EINECSNumber : 255-194-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only