missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 41052-75-9
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.04
MDL Number MFCD00012928
InChI Key ADODRSVGNHNKAT-UHFFFAOYSA-N
Synonym 2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride
PubChem CID 443287
ChEBI CHEBI:1046
IUPAC Name (2-chlorophenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC149140050
SDS View Documents
Thermo Scientific Chemicals
149140050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC149140250
SDS View Documents
Thermo Scientific Chemicals
149140250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

41052-75-9
179.04
ADODRSVGNHNKAT-UHFFFAOYSA-N
443287
(2-chlorophenyl)hydrazine;hydrochloride
C6H8Cl2N2
MFCD00012928
2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride
CHEBI:1046
[H+].[Cl-].NNC1=CC=CC=C1Cl

Specifications

41052-75-9
100.0
96% min. (Argentometry)
C6H8Cl2N2
MFCD00012928
15, 422
Solubility in water: soluble
[H+].[Cl-].NNC1=CC=CC=C1Cl
179.04
CHEBI:1046
97%
96.0
Authentic
Glass bottle
ClC6H4NHNH2·HCl
5g
2-chlorophenylhydrazine hydrochloride, 2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hydrochloride, 1-2-chlorophenyl hydrazine hydrochloride, o-chlorophenylhydrazine hcl, 2-chloro phenyl hydrazine hydrochloride, 2-chlorophenylhydrazine, chloride, hydrazine, 2-chlorophenyl-, monohydrochloride, 2-chlorophenylhydrazinehydrochloride
ADODRSVGNHNKAT-UHFFFAOYSA-N
(2-chlorophenyl)hydrazine;hydrochloride
443287
179.05
2-Chlorophenylhydrazine hydrochloride, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Do not eat,drink or smoke when using this product.
IF

GHS Signal Word: Warning

EINECSNumber : 255-194-1

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only