missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloropyridine-4-methanol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5009906

 View more versions of this product

Catalog No. AAH5009906


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloropyridine-4-methanol
64°C to 66°C
MFCD06858767
2-chloropyridin-4-yl methanol, 2-chloropyridine-4-methanol, 2-chloro-4-hydroxymethyl pyridine, 2-chloro-pyridine-4-yl-methanol, 2-chloro-pyridin-4-yl-methanol, 2-chloro-4-pyridinyl methanol, 4-pyridinemethanol, 2-chloro, 2-chloro-4-hydroxymethylpyridine, 2-chloro-4-pyridinemethanol, 2-chloro-4-pyridyl methan-1-ol
C1=CN=C(C=C1CO)Cl
143.57
143.57
100704-10-7
C6H6ClNO
5 g
UDDVPFLXGOBESH-UHFFFAOYSA-N
(2-chloropyridin-4-yl)methanol
7062237
97%

Chemical Identifiers

100704-10-7
143.57
UDDVPFLXGOBESH-UHFFFAOYSA-N
7062237
C1=CN=C(C=C1CO)Cl
C6H6ClNO
MFCD06858767
2-chloropyridin-4-yl methanol, 2-chloropyridine-4-methanol, 2-chloro-4-hydroxymethyl pyridine, 2-chloro-pyridine-4-yl-methanol, 2-chloro-pyridin-4-yl-methanol, 2-chloro-4-pyridinyl methanol, 4-pyridinemethanol, 2-chloro, 2-chloro-4-hydroxymethylpyridine, 2-chloro-4-pyridinemethanol, 2-chloro-4-pyridyl methan-1-ol
(2-chloropyridin-4-yl)methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only