Learn More
2-Chloropyridine-4-methanol, 97%, Thermo Scientific Chemicals
$1322.52 - $1322.52
Chemical Identifiers
CAS | 100704-10-7 |
---|---|
Molecular Formula | C6H6ClNO |
Molecular Weight (g/mol) | 143.57 |
MDL Number | MFCD06858767 |
InChI Key | UDDVPFLXGOBESH-UHFFFAOYSA-N |
Synonym | 2-chloropyridin-4-yl methanol, 2-chloropyridine-4-methanol, 2-chloro-4-hydroxymethyl pyridine, 2-chloro-pyridine-4-yl-methanol, 2-chloro-pyridin-4-yl-methanol, 2-chloro-4-pyridinyl methanol, 4-pyridinemethanol, 2-chloro, 2-chloro-4-hydroxymethylpyridine, 2-chloro-4-pyridinemethanol, 2-chloro-4-pyridyl methan-1-ol |
PubChem CID | 7062237 |
IUPAC Name | (2-chloropyridin-4-yl)methanol |
SMILES | C1=CN=C(C=C1CO)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5009906
|
Thermo Scientific Chemicals
H5009906 |
5 g |
Each for $1,322.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100704-10-7 | |
143.57 | |
UDDVPFLXGOBESH-UHFFFAOYSA-N | |
7062237 | |
C1=CN=C(C=C1CO)Cl |
C6H6ClNO | |
MFCD06858767 | |
2-chloropyridin-4-yl methanol, 2-chloropyridine-4-methanol, 2-chloro-4-hydroxymethyl pyridine, 2-chloro-pyridine-4-yl-methanol, 2-chloro-pyridin-4-yl-methanol, 2-chloro-4-pyridinyl methanol, 4-pyridinemethanol, 2-chloro, 2-chloro-4-hydroxymethylpyridine, 2-chloro-4-pyridinemethanol, 2-chloro-4-pyridyl methan-1-ol | |
(2-chloropyridin-4-yl)methanol |
Specifications
100704-10-7 | |
C6H6ClNO | |
5 g | |
UDDVPFLXGOBESH-UHFFFAOYSA-N | |
(2-chloropyridin-4-yl)methanol | |
7062237 | |
97% |
64°C to 66°C | |
MFCD06858767 | |
2-chloropyridin-4-yl methanol, 2-chloropyridine-4-methanol, 2-chloro-4-hydroxymethyl pyridine, 2-chloro-pyridine-4-yl-methanol, 2-chloro-pyridin-4-yl-methanol, 2-chloro-4-pyridinyl methanol, 4-pyridinemethanol, 2-chloro, 2-chloro-4-hydroxymethylpyridine, 2-chloro-4-pyridinemethanol, 2-chloro-4-pyridyl methan-1-ol | |
C1=CN=C(C=C1CO)Cl | |
143.57 | |
143.57 | |
2-Chloropyridine-4-methanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only