missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorothiobenzamide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1118014

 View more versions of this product

Catalog No. AAL1118014


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chlorothiobenzamide
15717-17-6
C7H6ClNS
508511
FLQYOORLPNYQEV-UHFFFAOYSA-N
2-chlorobenzenecarbothioamide
2734824
97%
63°C to 66°C
25 g
MFCD00040955
2-chlorothiobenzamide, 2-chlorobenzothioamide, benzenecarbothioamide, 2-chloro, 2-chlorobenzene-1-carbothioamide, o-chlorothiobenzamide, 2-chloro-thiobenzamide, acmc-1btqa, benzamide, o-chlorothio, 2-chloranylbenzenecarbothioamide, benzenecarbothioamide,2-chloro
C1=CC=C(C(=C1)C(=S)N)Cl
171.642
171.65

Chemical Identifiers

15717-17-6
171.642
FLQYOORLPNYQEV-UHFFFAOYSA-N
2-chlorobenzenecarbothioamide
C7H6ClNS
MFCD00040955
2734824
C1=CC=C(C(=C1)C(=S)N)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only