missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorothiobenzamide, 97%, Thermo Scientific Chemicals

$156.32 - $613.59

Chemical Identifiers

CAS 15717-17-6
Molecular Formula C7H6ClNS
Molecular Weight (g/mol) 171.642
MDL Number MFCD00040955
InChI Key FLQYOORLPNYQEV-UHFFFAOYSA-N
PubChem CID 2734824
IUPAC Name 2-chlorobenzenecarbothioamide
SMILES C1=CC=C(C(=C1)C(=S)N)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1118006
View Documents
Thermo Scientific Chemicals
L1118006
5 g
Each for $156.32
 
AAL1118014
View Documents
Thermo Scientific Chemicals
L1118014
25 g
Each for $613.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15717-17-6
171.642
FLQYOORLPNYQEV-UHFFFAOYSA-N
2-chlorobenzenecarbothioamide
C7H6ClNS
MFCD00040955
2734824
C1=CC=C(C(=C1)C(=S)N)Cl

Specifications

63°C to 66°C
5 g
MFCD00040955
2-chlorothiobenzamide, 2-chlorobenzothioamide, benzenecarbothioamide, 2-chloro, 2-chlorobenzene-1-carbothioamide, o-chlorothiobenzamide, 2-chloro-thiobenzamide, acmc-1btqa, benzamide, o-chlorothio, 2-chloranylbenzenecarbothioamide, benzenecarbothioamide,2-chloro
C1=CC=C(C(=C1)C(=S)N)Cl
171.642
171.65
2-Chlorothiobenzamide
15717-17-6
C7H6ClNS
508511
FLQYOORLPNYQEV-UHFFFAOYSA-N
2-chlorobenzenecarbothioamide
2734824
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only