missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyclohexylethanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2078230

 View more versions of this product

Catalog No. AAB2078230

Explore available promotions

Description

Description

Used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent.

Solubility
Soluble in alcohol and dipropylene glycol. Soluble in water (1750 mg/L at 25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2-Cyclohexylethanol
-20°C
206°C to 207°C
C8H16O
MFCD00001525
1848152
Soluble in alcohol and dipropylene glycol. Soluble in water (1750mg/L at 25°C).
C1CCC(CC1)CCO
128.215
128.22
4442-79-9
0.92
86°C (186°F)
1.466
250 g
cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol
QJQZRLXDLORINA-UHFFFAOYSA-N
2-cyclohexylethanol
20508
99%

Chemical Identifiers

4442-79-9
128.215
QJQZRLXDLORINA-UHFFFAOYSA-N
20508
C1CCC(CC1)CCO
C8H16O
MFCD00001525
cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol
2-cyclohexylethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H227
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

EINECSNumber : 224-672-1

RTECSNumber : KK3528000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only