Learn More
2-Cyclohexylethanol, 99%
CAS: 4442-79-9 | C8H16O | 128.215 g/mol
$98.33 - $804.97
Chemical Identifiers
| CAS | 4442-79-9 |
|---|---|
| Molecular Formula | C8H16O |
| Molecular Weight (g/mol) | 128.215 |
| MDL Number | MFCD00001525 |
| InChI Key | QJQZRLXDLORINA-UHFFFAOYSA-N |
| Synonym | cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol |
| PubChem CID | 20508 |
| IUPAC Name | 2-cyclohexylethanol |
| SMILES | C1CCC(CC1)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2078218
|
Thermo Scientific Chemicals
B2078218 |
50 g |
Each for $98.33
|
|
|||||
|
AAB2078230
|
Thermo Scientific Chemicals
B2078230 |
250 g |
Each for $255.97
|
|
|||||
|
AAB207820B
|
Thermo Scientific Chemicals
B207820B |
1000 g |
Each for $804.97
|
|
|||||
Description
Used as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a flavoring agent.
Solubility
Soluble in alcohol and dipropylene glycol. Soluble in water (1750 mg/L at 25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 4442-79-9 | |
| 128.215 | |
| QJQZRLXDLORINA-UHFFFAOYSA-N | |
| 20508 | |
| C1CCC(CC1)CCO |
| C8H16O | |
| MFCD00001525 | |
| cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol | |
| 2-cyclohexylethanol |
Specifications
| 4442-79-9 | |
| 0.92 | |
| 86°C (186°F) | |
| 1.466 | |
| 50 g | |
| cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol | |
| QJQZRLXDLORINA-UHFFFAOYSA-N | |
| 2-cyclohexylethanol | |
| 20508 | |
| 99% |
| -20°C | |
| 206°C to 207°C | |
| C8H16O | |
| MFCD00001525 | |
| 1848152 | |
| Soluble in alcohol and dipropylene glycol. Soluble in water (1750mg/L at 25°C). | |
| C1CCC(CC1)CCO | |
| 128.215 | |
| 128.22 | |
| 2-Cyclohexylethanol |
Safety and Handling
GHS H Statement
H302-H312-H227
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
EINECSNumber : 224-672-1
RTECSNumber : KK3528000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only