missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1108003

 View more versions of this product

Catalog No. AAL1108003



Description

Description

2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

2-Cyclopropylethanol
137°C to 138°C
C5H10O
MFCD00040762
UN1987
cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687
LUNMJRJMSXZSLC-UHFFFAOYSA-N
2-cyclopropylethanol
137642
96%
2566-44-1
47°C (116°F)
1.4355
1 g
2036028
Miscible with water.
OCCC1CC1
86.13
86.13

Chemical Identifiers

2566-44-1
86.13
LUNMJRJMSXZSLC-UHFFFAOYSA-N
137642
C5H10O
MFCD00040762
cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687
OCCC1CC1
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only