Learn More
2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals
$83.63 - $695.01
Chemical Identifiers
CAS | 2566-44-1 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00040762 |
InChI Key | LUNMJRJMSXZSLC-UHFFFAOYSA-N |
Synonym | cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687 |
PubChem CID | 137642 |
SMILES | OCCC1CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL11080MD
|
Thermo Scientific Chemicals
L11080MD |
250 mg |
Each for $83.63
|
|
|||||
AAL1108003
|
Thermo Scientific Chemicals
L1108003 |
1 g |
Each for $207.45
|
|
|||||
AAL1108006
|
Thermo Scientific Chemicals
L1108006 |
5 g |
Each for $695.01
|
|
|||||
Description
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
2566-44-1 | |
86.13 | |
LUNMJRJMSXZSLC-UHFFFAOYSA-N | |
137642 |
C5H10O | |
MFCD00040762 | |
cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687 | |
OCCC1CC1 |
Specifications
2566-44-1 | |
47°C (116°F) | |
1.4355 | |
250 mg | |
2036028 | |
Miscible with water. | |
OCCC1CC1 | |
86.13 | |
86.13 | |
2-Cyclopropylethanol |
137°C to 138°C | |
C5H10O | |
MFCD00040762 | |
UN1987 | |
cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687 | |
LUNMJRJMSXZSLC-UHFFFAOYSA-N | |
2-cyclopropylethanol | |
137642 | |
96% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only