missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 114032500

 View more versions of this product

Catalog No. AC114032500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Diethylaminoethylchloride hydrochloride
99.4
208.0°C to 212.0°C
1% max. (105°C, 3 hrs)
99.5%
C6H15Cl2N
MFCD00012519
04, III, 240
Solubility in water: 2000g/L (20°C)
[H+].[Cl-].CCN(CC)CCCl
172.09
172.1
Crystalline Powder
869-24-9
100.0
White to Beige
Authentic
Glass Bottle
(C2H5)2NCH2CH2Cl·HCl
250 g
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
RAGSWDIQBBZLLL-UHFFFAOYSA-N
2-chloro-N,N-diethylethanamine;hydrochloride
13363
99.5%

Chemical Identifiers

869-24-9
172.09
RAGSWDIQBBZLLL-UHFFFAOYSA-N
13363
[H+].[Cl-].CCN(CC)CCCl
C6H15Cl2N
MFCD00012519
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
2-chloro-N,N-diethylethanamine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 212-786-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only