missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals

$49.59 - $458.83

Chemical Identifiers

CAS 869-24-9
Molecular Formula C6H15Cl2N
Molecular Weight (g/mol) 172.09
MDL Number MFCD00012519
InChI Key RAGSWDIQBBZLLL-UHFFFAOYSA-N
Synonym 2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
PubChem CID 13363
IUPAC Name 2-chloro-N,N-diethylethanamine;hydrochloride
SMILES [H+].[Cl-].CCN(CC)CCCl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC114030050
SDS View Documents
Thermo Scientific Chemicals
114030050
5 g Glass Bottle
Each for $49.59
 
AC114032500
SDS View Documents
Thermo Scientific Chemicals
114032500
250 g Glass Bottle
Each for $73.75
 
AC114030010
SDS View Documents
Thermo Scientific Chemicals
114030010
1 kg Glass Bottle
Each for $216.05
 
AC114030025
SDS View Documents
Thermo Scientific Chemicals
114030025
2.5 kg Plastic drum
Each for $458.83
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

869-24-9
172.09
RAGSWDIQBBZLLL-UHFFFAOYSA-N
13363
[H+].[Cl-].CCN(CC)CCCl
C6H15Cl2N
MFCD00012519
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
2-chloro-N,N-diethylethanamine;hydrochloride

Specifications

869-24-9
100.0
White to Beige
Authentic
Glass Bottle
(C2H5)2NCH2CH2Cl·HCl
5 g
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
RAGSWDIQBBZLLL-UHFFFAOYSA-N
2-chloro-N,N-diethylethanamine;hydrochloride
13363
99.5%
2-Diethylaminoethylchloride hydrochloride
99.4
208.0°C to 212.0°C
1% max. (105°C, 3 hrs)
99.5%
C6H15Cl2N
MFCD00012519
04, III, 240
Solubility in water: 2000g/L (20°C)
[H+].[Cl-].CCN(CC)CCCl
172.09
172.1
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 212-786-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only