missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 156020250

 View more versions of this product

Catalog No. AC156020250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Ethoxyethanol
98.5
-70°C
135.0°C
Authentic
Metal drum
1.4060 to 1.4100
MFCD00002869
01, 467
15, 3803
Solubility in water: complete solubility. Other solubilities: miscible with most organic solvents
CCOCCO
90.12
2.08 mPa.s (20°C)
90.12
Extra Pure
110-80-5
100.0
0.9300g/mL
40°C
98.5% min. (GC)
C4H10O2
C2H5OCH2CH2OH
25 L
0.93
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
2-ethoxyethanol
8076
CHEBI:46788
99%
Liquid

Chemical Identifiers

110-80-5
90.12
ZNQVEEAIQZEUHB-UHFFFAOYSA-N
8076
2-ethoxyethanol
C4H10O2
MFCD00002869
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
CHEBI:46788
CCOCCO
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
May damage fertility.
May damage the unborn child.
Toxic if inhaled.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 203-804-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only