Learn More
2-Ethoxyethanol, 99%, extra pure, Thermo Scientific Chemicals
$122.45 - $1212.47
Chemical Identifiers
CAS | 110-80-5 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
MDL Number | MFCD00002869 |
InChI Key | ZNQVEEAIQZEUHB-UHFFFAOYSA-N |
Synonym | cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee |
PubChem CID | 8076 |
ChEBI | CHEBI:46788 |
IUPAC Name | 2-ethoxyethanol |
SMILES | CCOCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC156020010
|
Thermo Scientific Chemicals
156020010 |
1 L | Glass Bottle |
Each for $122.45
|
|
||||
AC156020025
|
Thermo Scientific Chemicals
156020025 |
2.5 L | Glass Bottle |
Each for $243.78
Case of 4 Each for $0.00
|
|
||||
AC156020250
|
Thermo Scientific Chemicals
156020250 |
25 L | Metal drum |
Each for $1,212.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-80-5 | |
90.12 | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
8076 | |
2-ethoxyethanol |
C4H10O2 | |
MFCD00002869 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
CHEBI:46788 | |
CCOCCO |
Specifications
110-80-5 | |
100.0 | |
0.9300g/mL | |
40°C | |
98.5% min. (GC) | |
C4H10O2 | |
C2H5OCH2CH2OH | |
1 L | |
0.93 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
2-ethoxyethanol | |
8076 | |
CHEBI:46788 | |
99% | |
Liquid |
98.5 | |
-70°C | |
135.0°C | |
Authentic | |
Glass Bottle | |
1.4060 to 1.4100 | |
MFCD00002869 | |
01, 467 | |
15, 3803 | |
Solubility in water: complete solubility. Other solubilities: miscible with most organic solvents | |
CCOCCO | |
90.12 | |
2.08 mPa.s (20°C) | |
90.12 | |
Extra Pure | |
2-Ethoxyethanol |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May damage fertility.
May damage the unborn child.
Toxic if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 203-804-1
RUO – Research Use Only