Learn More
2-Ethylbenzoic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 378750010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Ethylbenzoic acid | |
96.0 | |
Authentic | |
Glass bottle | |
CH3CH2C6H4COOH | |
1g | |
CGMMPMYKMDITEA-UHFFFAOYSA-N | |
2-ethylbenzoic acid | |
34170 | |
97% |
612-19-1 | |
100.0 | |
96% min. (HPLC) | |
C9H10O2 | |
MFCD00045838 | |
benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid | |
CCC1=CC=CC=C1C(=O)O | |
150.18 | |
150.18 |
Chemical Identifiers
612-19-1 | |
150.18 | |
CGMMPMYKMDITEA-UHFFFAOYSA-N | |
34170 | |
CCC1=CC=CC=C1C(=O)O |
C9H10O2 | |
MFCD00045838 | |
benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid | |
2-ethylbenzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO â Research Use Only