missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbenzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 612-19-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00045838
InChI Key CGMMPMYKMDITEA-UHFFFAOYSA-N
Synonym benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid
PubChem CID 34170
IUPAC Name 2-ethylbenzoic acid
SMILES CCC1=CC=CC=C1C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC378750010
SDS View Documents
Thermo Scientific Chemicals
378750010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC378750050
View Documents
Thermo Scientific Chemicals
378750050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

612-19-1
150.18
CGMMPMYKMDITEA-UHFFFAOYSA-N
34170
CCC1=CC=CC=C1C(=O)O
C9H10O2
MFCD00045838
benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid
2-ethylbenzoic acid

Specifications

612-19-1
100.0
96% min. (HPLC)
C9H10O2
MFCD00045838
benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid
CCC1=CC=CC=C1C(=O)O
150.18
150.18
2-Ethylbenzoic acid
96.0
Authentic
Glass bottle
CH3CH2C6H4COOH
1g
CGMMPMYKMDITEA-UHFFFAOYSA-N
2-ethylbenzoic acid
34170
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only