missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylbenzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 378750050

 View more versions of this product

Catalog No. AC378750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Ethylbenzoic acid
96.0
Authentic
Glass bottle
CH3CH2C6H4COOH
5g
CGMMPMYKMDITEA-UHFFFAOYSA-N
2-ethylbenzoic acid
34170
97%
612-19-1
100.0
96% min. (HPLC)
C9H10O2
MFCD00045838
benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid
CCC1=CC=CC=C1C(=O)O
150.18
150.18

Chemical Identifiers

612-19-1
150.18
CGMMPMYKMDITEA-UHFFFAOYSA-N
34170
CCC1=CC=CC=C1C(=O)O
C9H10O2
MFCD00045838
benzoic acid, ethyl, o-ethylbenzoic acid, 2-ethylbenzoicacidethylester, ethylbenzoic acid, pubchem9869, 2-ethyl-benzoic acid, benzoic acid,2-ethyl, benzoic acid, 2-ethyl, acmc-1ax4g, 2-ethylbenzoic acid
2-ethylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only