missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethylcyclohexanol (cis- and trans- mixture) 97.0+%, TCI America™
Supplier: TCI America E0365500ML
Specifications
2-Ethylcyclohexanol (cis- and trans- mixture) | |
Colorless | |
C8H16O | |
500 mL | |
CCC1CCCCC1O | |
128.215 | |
128.22 | |
Liquid |
3760-20-1 | |
79°C | |
MFCD00001445 | |
CFYUBZHJDXXXQE-UHFFFAOYSA-N | |
2-ethylcyclohexan-1-ol | |
19576 | |
≥97.0% (GC) |
Chemical Identifiers
3760-20-1 | |
128.215 | |
CFYUBZHJDXXXQE-UHFFFAOYSA-N | |
2-ethylcyclohexan-1-ol |
C8H16O | |
MFCD00001445 | |
19576 | |
CCC1CCCCC1O |
Safety and Handling
TSCA : No