missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylcyclohexanol (cis- and trans- mixture) 97.0+%, TCI America™

$213.30 - $926.27

Chemical Identifiers

CAS 3760-20-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001445
InChI Key CFYUBZHJDXXXQE-UHFFFAOYSA-N
PubChem CID 19576
IUPAC Name 2-ethylcyclohexan-1-ol
SMILES CCC1CCCCC1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E036525ML
View Documents
TCI America
E036525ML
25 mL
Each for $213.30
 
E0365500ML
View Documents
TCI America
E0365500ML
500 mL
Each for $926.27
 
Description

Description

Specifications

Chemical Identifiers

3760-20-1
128.215
CFYUBZHJDXXXQE-UHFFFAOYSA-N
2-ethylcyclohexan-1-ol
C8H16O
MFCD00001445
19576
CCC1CCCCC1O

Specifications

3760-20-1
79°C
MFCD00001445
CFYUBZHJDXXXQE-UHFFFAOYSA-N
2-ethylcyclohexan-1-ol
19576
≥97.0% (GC)
2-Ethylcyclohexanol (cis- and trans- mixture)
Colorless
C8H16O
25 mL
CCC1CCCCC1O
128.215
128.22
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'