missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6323506

 View more versions of this product

Catalog No. AAH6323506



Description

Description

2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

2-Fluoro-3-nitrobenzoic acid
C7H4FNO4
5 g
Slightly soluble in water.
OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O
185.11
185.11
Solid
317-46-4
MFCD06797825
2-fluoro-3-nitrobenzoic acid, 2-fluoro-3-nitrobenzoicacid, 2-fluoro-3-nitro benzoic acid, 3-carboxy-2-fluoronitrobenzene, pubchem1398, acmc-1ajf3, ksc494k8n, benzoic acid,2-fluoro-3-nitro
WLGUSLGYTNJJFV-UHFFFAOYSA-N
2-fluoro-3-nitrobenzoic acid
18672992
97%

Chemical Identifiers

317-46-4
185.11
WLGUSLGYTNJJFV-UHFFFAOYSA-N
18672992
OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O
C7H4FNO4
MFCD06797825
2-fluoro-3-nitrobenzoic acid, 2-fluoro-3-nitrobenzoicacid, 2-fluoro-3-nitro benzoic acid, 3-carboxy-2-fluoronitrobenzene, pubchem1398, acmc-1ajf3, ksc494k8n, benzoic acid,2-fluoro-3-nitro
2-fluoro-3-nitrobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only