Learn More
2-Fluoro-3-nitrobenzoic acid, 97%
CAS: 317-46-4 | C7H4FNO4 | 185.11 g/mol
$70.08 - $234.78
Chemical Identifiers
| CAS | 317-46-4 |
|---|---|
| Molecular Formula | C7H4FNO4 |
| Molecular Weight (g/mol) | 185.11 |
| MDL Number | MFCD06797825 |
| InChI Key | WLGUSLGYTNJJFV-UHFFFAOYSA-N |
| Synonym | 2-fluoro-3-nitrobenzoic acid, 2-fluoro-3-nitrobenzoicacid, 2-fluoro-3-nitro benzoic acid, 3-carboxy-2-fluoronitrobenzene, pubchem1398, acmc-1ajf3, ksc494k8n, benzoic acid,2-fluoro-3-nitro |
| PubChem CID | 18672992 |
| IUPAC Name | 2-fluoro-3-nitrobenzoic acid |
| SMILES | OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6323503
|
Thermo Scientific Chemicals
H6323503 |
1 g |
Each for $70.08
|
|
|||||
|
AAH6323506
|
Thermo Scientific Chemicals
H6323506 |
5 g |
Each for $234.78
|
|
|||||
Description
2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 317-46-4 | |
| 185.11 | |
| WLGUSLGYTNJJFV-UHFFFAOYSA-N | |
| 18672992 | |
| OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O |
| C7H4FNO4 | |
| MFCD06797825 | |
| 2-fluoro-3-nitrobenzoic acid, 2-fluoro-3-nitrobenzoicacid, 2-fluoro-3-nitro benzoic acid, 3-carboxy-2-fluoronitrobenzene, pubchem1398, acmc-1ajf3, ksc494k8n, benzoic acid,2-fluoro-3-nitro | |
| 2-fluoro-3-nitrobenzoic acid |
Specifications
| 317-46-4 | |
| MFCD06797825 | |
| 2-fluoro-3-nitrobenzoic acid, 2-fluoro-3-nitrobenzoicacid, 2-fluoro-3-nitro benzoic acid, 3-carboxy-2-fluoronitrobenzene, pubchem1398, acmc-1ajf3, ksc494k8n, benzoic acid,2-fluoro-3-nitro | |
| WLGUSLGYTNJJFV-UHFFFAOYSA-N | |
| 2-fluoro-3-nitrobenzoic acid | |
| 18672992 | |
| 97% | |
| 2-Fluoro-3-nitrobenzoic acid |
| C7H4FNO4 | |
| 1 g | |
| Slightly soluble in water. | |
| OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O | |
| 185.11 | |
| 185.11 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only