missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

2-Fluoro-4-methoxybenzonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H3253106

 Afficher plus de versions de ce produit

Numéro de catalogue. AAH3253106


Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-4-methoxybenzonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Spécifications

Spécifications

2-Fluoro-4-methoxybenzonitrile
62°C to 65°C
MFCD00509374
UN3439
HWKUZTFIZATJPM-UHFFFAOYSA-N
2-fluoro-4-methoxybenzonitrile
639878
97%
94610-82-9
C8H6FNO
5 g
3-fluoro-4-cyanoanisole, 2-fluoro-4-methoxy-benzonitrile, benzonitrile, 2-fluoro-4-methoxy, 3-fluoro-4-cyano anisole, 4-cyano-3-fluoroanisole, buttpark 81\01-98, 2-fluoro-4-methoxybenzenecarbonitrile, pubchem4783, 2-fluoro-4-methoxybe
COC1=CC(F)=C(C=C1)C#N
151.14
151.14

Identifiants chimiques

94610-82-9
151.14
HWKUZTFIZATJPM-UHFFFAOYSA-N
639878
COC1=CC(F)=C(C=C1)C#N
C8H6FNO
MFCD00509374
3-fluoro-4-cyanoanisole, 2-fluoro-4-methoxy-benzonitrile, benzonitrile, 2-fluoro-4-methoxy, 3-fluoro-4-cyano anisole, 4-cyano-3-fluoroanisole, buttpark 81\01-98, 2-fluoro-4-methoxybenzenecarbonitrile, pubchem4783, 2-fluoro-4-methoxybe
2-fluoro-4-methoxybenzonitrile
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Offres spéciales

Offres spéciales