missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxyphenylacetonitrile, 97%

CAS: 749934-29-0 | C9H8FNO | 165.17 g/mol

Supplier:  Thermo Scientific Chemicals H30752MD

 View more versions of this product

Catalog No. AAH30752MD

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluoro-4-methoxyphenylacetonitrile
C9H8FNO
MFCD09038467
UN3276
OKPZYRPNHFQVMZ-UHFFFAOYSA-N
2-(2-fluoro-4-methoxyphenyl)acetonitrile
26343860
97%
749934-29-0
1.5131
250 mg
4-methoxy-2-fluorobenzyl cyanide, 2-2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxyphenylacetonitrile, 2-fluoro-4-methoxybenzyl cyanide, 2-fluoro-4-methoxybenzeneacetonitrile, 2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxy-phenyl-acetonitrile, 2-2-fluoro-4-methoxyphenyl ethanenitrile, 2-2-fluoranyl-4-methoxy-phenyl ethanenitrile, benzeneacetonitrile, 2-fluoro-4-methoxy-9ci
COC1=CC(F)=C(CC#N)C=C1
165.17
165.17

Chemical Identifiers

749934-29-0
165.17
OKPZYRPNHFQVMZ-UHFFFAOYSA-N
26343860
COC1=CC(F)=C(CC#N)C=C1
C9H8FNO
MFCD09038467
4-methoxy-2-fluorobenzyl cyanide, 2-2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxyphenylacetonitrile, 2-fluoro-4-methoxybenzyl cyanide, 2-fluoro-4-methoxybenzeneacetonitrile, 2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxy-phenyl-acetonitrile, 2-2-fluoro-4-methoxyphenyl ethanenitrile, 2-2-fluoranyl-4-methoxy-phenyl ethanenitrile, benzeneacetonitrile, 2-fluoro-4-methoxy-9ci
2-(2-fluoro-4-methoxyphenyl)acetonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only