missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-methoxyphenylacetonitrile, 97%, Thermo Scientific Chemicals

$141.36 - $199.05

Chemical Identifiers

CAS 749934-29-0
Molecular Formula C9H8FNO
Molecular Weight (g/mol) 165.17
MDL Number MFCD09038467
InChI Key OKPZYRPNHFQVMZ-UHFFFAOYSA-N
Synonym 4-methoxy-2-fluorobenzyl cyanide, 2-2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxyphenylacetonitrile, 2-fluoro-4-methoxybenzyl cyanide, 2-fluoro-4-methoxybenzeneacetonitrile, 2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxy-phenyl-acetonitrile, 2-2-fluoro-4-methoxyphenyl ethanenitrile, 2-2-fluoranyl-4-methoxy-phenyl ethanenitrile, benzeneacetonitrile, 2-fluoro-4-methoxy-9ci
PubChem CID 26343860
IUPAC Name 2-(2-fluoro-4-methoxyphenyl)acetonitrile
SMILES COC1=CC(F)=C(CC#N)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH30752MD
View Documents
Thermo Scientific Chemicals
H30752MD
250 mg
Each for $141.36
 
AAH3075203
View Documents
Thermo Scientific Chemicals
H3075203
1 g
Each for $199.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

749934-29-0
165.17
OKPZYRPNHFQVMZ-UHFFFAOYSA-N
26343860
COC1=CC(F)=C(CC#N)C=C1
C9H8FNO
MFCD09038467
4-methoxy-2-fluorobenzyl cyanide, 2-2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxyphenylacetonitrile, 2-fluoro-4-methoxybenzyl cyanide, 2-fluoro-4-methoxybenzeneacetonitrile, 2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxy-phenyl-acetonitrile, 2-2-fluoro-4-methoxyphenyl ethanenitrile, 2-2-fluoranyl-4-methoxy-phenyl ethanenitrile, benzeneacetonitrile, 2-fluoro-4-methoxy-9ci
2-(2-fluoro-4-methoxyphenyl)acetonitrile

Specifications

749934-29-0
1.5131
250 mg
4-methoxy-2-fluorobenzyl cyanide, 2-2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxyphenylacetonitrile, 2-fluoro-4-methoxybenzyl cyanide, 2-fluoro-4-methoxybenzeneacetonitrile, 2-fluoro-4-methoxyphenyl acetonitrile, 2-fluoro-4-methoxy-phenyl-acetonitrile, 2-2-fluoro-4-methoxyphenyl ethanenitrile, 2-2-fluoranyl-4-methoxy-phenyl ethanenitrile, benzeneacetonitrile, 2-fluoro-4-methoxy-9ci
COC1=CC(F)=C(CC#N)C=C1
165.17
165.17
2-Fluoro-4-methoxyphenylacetonitrile
C9H8FNO
MFCD09038467
UN3276
OKPZYRPNHFQVMZ-UHFFFAOYSA-N
2-(2-fluoro-4-methoxyphenyl)acetonitrile
26343860
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only