missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2563003

 View more versions of this product

Catalog No. AAB2563003


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
1.418
70°C (158°F)
1.455
1 g
Moisture sensitive
NAIKHCBDZGSGHH-UHFFFAOYSA-N
1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene
2733380
97%
69922-27-6
53°C (2.5 mmHg)
C8H3F4NO
MFCD00673069
UN2206
2-fluoro-5-trifluoromethyl phenyl isocyanate, 1-fluoro-2-isocyanato-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl phenylisocyanate, 2-fluoro-5-trifluoromethylphenyl isocyanate, benzene,1-fluoro-2-isocyanato-4-trifluoromethyl, benzene, 1-fluoro-2-isocyanato-4-trifluoromethyl, acmc-20anuz, pubchem12480, 2-fluoro-5-trifluoromethylphenylisocyanate, 2-fluoro-5-trifluoromethyl phenyl isocyan
FC1=CC=C(C=C1N=C=O)C(F)(F)F
205.11
205.11

Chemical Identifiers

69922-27-6
205.11
NAIKHCBDZGSGHH-UHFFFAOYSA-N
1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene
C8H3F4NO
MFCD00673069
2733380
FC1=CC=C(C=C1N=C=O)C(F)(F)F
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only