Learn More
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
$161.19 - $579.14
Chemical Identifiers
CAS | 69922-27-6 |
---|---|
Molecular Formula | C8H3F4NO |
Molecular Weight (g/mol) | 205.11 |
MDL Number | MFCD00673069 |
InChI Key | NAIKHCBDZGSGHH-UHFFFAOYSA-N |
PubChem CID | 2733380 |
IUPAC Name | 1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene |
SMILES | FC1=CC=C(C=C1N=C=O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2563003
|
Thermo Scientific Chemicals
B2563003 |
1 g |
Each for $161.19
|
|
|||||
AAB2563006
|
Thermo Scientific Chemicals
B2563006 |
5 g |
Each for $579.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69922-27-6 | |
205.11 | |
NAIKHCBDZGSGHH-UHFFFAOYSA-N | |
1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene |
C8H3F4NO | |
MFCD00673069 | |
2733380 | |
FC1=CC=C(C=C1N=C=O)C(F)(F)F |
Specifications
69922-27-6 | |
53°C (2.5 mmHg) | |
C8H3F4NO | |
MFCD00673069 | |
UN2206 | |
2-fluoro-5-trifluoromethyl phenyl isocyanate, 1-fluoro-2-isocyanato-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl phenylisocyanate, 2-fluoro-5-trifluoromethylphenyl isocyanate, benzene,1-fluoro-2-isocyanato-4-trifluoromethyl, benzene, 1-fluoro-2-isocyanato-4-trifluoromethyl, acmc-20anuz, pubchem12480, 2-fluoro-5-trifluoromethylphenylisocyanate, 2-fluoro-5-trifluoromethyl phenyl isocyan | |
FC1=CC=C(C=C1N=C=O)C(F)(F)F | |
205.11 | |
205.11 | |
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate |
1.418 | |
70°C (158°F) | |
1.455 | |
1 g | |
Moisture sensitive | |
NAIKHCBDZGSGHH-UHFFFAOYSA-N | |
1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene | |
2733380 | |
97% |
Safety and Handling
GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only