missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-m-xylene, 98%, Thermo Scientific Chemicals

$140.52 - $439.08

Chemical Identifiers

CAS 443-88-9
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.158
MDL Number MFCD00039215
InChI Key JTAUTNBVFDTYTI-UHFFFAOYSA-N
Synonym 2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
PubChem CID 123065
IUPAC Name 2-fluoro-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1042406
View Documents
Thermo Scientific Chemicals
A1042406
5 g
Each for $140.52
 
AAA1042414
View Documents
Thermo Scientific Chemicals
A1042414
25 g
Each for $439.08
 
Description

Description

It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Specifications

Chemical Identifiers

443-88-9
124.158
JTAUTNBVFDTYTI-UHFFFAOYSA-N
123065
CC1=C(C(=CC=C1)C)F
C8H9F
MFCD00039215
2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
2-fluoro-1,3-dimethylbenzene

Specifications

443-88-9
147°C to 148°C
Aromatic
C8H9F
UN1993
2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
JTAUTNBVFDTYTI-UHFFFAOYSA-N
2-fluoro-1,3-dimethylbenzene
123065
98%
0.988
5 g
1.479
MFCD00039215
1857413
Not miscible or difficult to mix in water.
CC1=C(C(=CC=C1)C)F
124.158
124.16
2-Fluoro-m-xylene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 207-144-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only