missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Furoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 119820050

 View more versions of this product

Catalog No. AC119820050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Furoic acid
88-14-2
100.0
Beige to White
137°C
98%
C5H4O3
5 g
15, 4336
Solubility in water: 36g/L (20°C). Other solubilities: soluble in alcohol and ether,insoluble in paraffin hydrocarbons
C1=COC(=C1)C(=O)O
112.08
CHEBI:30845
98%
98%
97.5
129.0°C to 133.0°C
230.0°C to 232.0°C
Authentic
Glass bottle
MFCD00003238
18, 272
2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
furan-2-carboxylic acid
6919
112.08
Fine Crystalline Powder

Chemical Identifiers

88-14-2
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
6919
furan-2-carboxylic acid
MFCD00003238
2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
CHEBI:30845
C1=COC(=C1)C(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 201-803-0

RTECSNumber : LV1763000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only