missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Furoic acid, 98%, Thermo Scientific Chemicals

$39.94 - $72.66

Chemical Identifiers

CAS 88-14-2
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119820050
SDS View Documents
Thermo Scientific Chemicals
119820050
5 g Glass bottle
Each for $39.94
 
AC119821000
SDS View Documents
Thermo Scientific Chemicals
119821000
100 g Plastic bottle
Each for $72.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

88-14-2
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
6919
furan-2-carboxylic acid
MFCD00003238
2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
CHEBI:30845
C1=COC(=C1)C(=O)O

Specifications

88-14-2
100.0
Beige to White
137°C
98%
C5H4O3
5 g
15, 4336
Solubility in water: 36g/L (20°C). Other solubilities: soluble in alcohol and ether,insoluble in paraffin hydrocarbons
C1=COC(=C1)C(=O)O
112.08
CHEBI:30845
98%
2-Furoic acid, 98%
97.5
129.0°C to 133.0°C
230.0°C to 232.0°C
Authentic
Glass bottle
MFCD00003238
18, 272
2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
furan-2-carboxylic acid
6919
112.08
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 201-803-0

RTECSNumber : LV1763000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only