Learn More
2-Hexanone, 98%, Thermo Scientific Chemicals
2-Hexanone, 98%, C6H12O, CAS Number-591-78-6, 2-hexanone, 2-oxohexane, butyl methyl ketone, hexanone, hexanone-2, ketone, butyl methyl, methyl butyl ketone, methyl n-butyl ketone, n-butyl methyl ketone, propylacetone, 500mL, 01, 689, 127 deg.C, 100.0, 97.5, Colorless to Yellow, 0.8100g/mL, 209-731-1
Supplier: Thermo Scientific Chemicals 146881000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Hexanone | |
591-78-6 | |
100.0 | |
Colorless to Yellow | |
127°C | |
Authentic | |
Glass bottle | |
1.3995 to 1.4015 | |
MFCD00009482 | |
01, 689 | |
15, 6107 | |
Solubility in water: 20g/l (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCC(C)=O | |
100.16 | |
100.16 | |
Liquid |
98% | |
97.5 | |
-57°C | |
0.8100g/mL | |
23°C | |
98% | |
C6H12O | |
CH3(CH2)3COCH3 | |
100 mL | |
0.81 | |
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl | |
QQZOPKMRPOGIEB-UHFFFAOYSA-N | |
hexan-2-one | |
11583 | |
98% |
Chemical Identifiers
591-78-6 | |
100.16 | |
QQZOPKMRPOGIEB-UHFFFAOYSA-N | |
11583 | |
CCCCC(C)=O |
C6H12O | |
MFCD00009482 | |
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl | |
hexan-2-one |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Flammable liquid and vapor.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-731-1
RTECSNumber : MP1400000
TSCA : TSCA
RUO – Research Use Only