missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, 98%

2-Hexanone, 98%, C6H12O, CAS Number-591-78-6, 2-hexanone, 2-oxohexane, butyl methyl ketone, hexanone, hexanone-2, ketone, butyl methyl, methyl butyl ketone, methyl n-butyl ketone, n-butyl methyl ketone, propylacetone, 500mL, 01, 689, 127 deg.C, 100.0, 97.5, Colorless to Yellow, 0.8100g/mL, 209-731-1 | CAS: 591-78-6 | C6H12O | 100.16 g/mol

$250.65 - $617.69

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC146881000
SDS View Documents
Thermo Scientific Chemicals
146881000
100 mL Glass bottle
Each for $250.65
Only null left
 
AC146885000
SDS View Documents
Thermo Scientific Chemicals
146885000
500 mL Glass bottle
Each for $617.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

591-78-6
100.16
QQZOPKMRPOGIEB-UHFFFAOYSA-N
11583
CCCCC(C)=O
C6H12O
MFCD00009482
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
hexan-2-one

Specifications

591-78-6
100.0
Colorless to Yellow
127°C
Authentic
Glass bottle
1.3995 to 1.4015
MFCD00009482
01, 689
15, 6107
Solubility in water: 20g/l (20°C). Other solubilities: soluble in alcohol and ether
CCCCC(C)=O
100.16
100.16
Liquid
97.5
-57°C
0.8100g/mL
23°C
98%
C6H12O
CH3(CH2)3COCH3
100 mL
0.81
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
QQZOPKMRPOGIEB-UHFFFAOYSA-N
hexan-2-one
11583
98%
2-Hexanone, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Flammable liquid and vapor.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-731-1

RTECSNumber : MP1400000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only