missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hexanone, 98%, Thermo Scientific Chemicals

2-Hexanone, 98%, C6H12O, CAS Number-591-78-6, 2-hexanone, 2-oxohexane, butyl methyl ketone, hexanone, hexanone-2, ketone, butyl methyl, methyl butyl ketone, methyl n-butyl ketone, n-butyl methyl ketone, propylacetone, 500mL, 01, 689, 127 deg.C, 100.0, 97.5, Colorless to Yellow, 0.8100g/mL, 209-731-1

Supplier:  Thermo Scientific Chemicals 146885000

 View more versions of this product

Catalog No. AC146885000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Hexanone
591-78-6
100.0
Colorless to Yellow
127°C
Authentic
Glass bottle
1.3995 to 1.4015
MFCD00009482
01, 689
15, 6107
Solubility in water: 20g/l (20°C). Other solubilities: soluble in alcohol and ether
CCCCC(C)=O
100.16
100.16
Liquid
98%
97.5
-57°C
0.8100g/mL
23°C
98%
C6H12O
CH3(CH2)3COCH3
500 mL
0.81
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
QQZOPKMRPOGIEB-UHFFFAOYSA-N
hexan-2-one
11583
98%

Chemical Identifiers

591-78-6
100.16
QQZOPKMRPOGIEB-UHFFFAOYSA-N
11583
CCCCC(C)=O
C6H12O
MFCD00009482
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
hexan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Flammable liquid and vapor.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-731-1

RTECSNumber : MP1400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only