missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1458503

 View more versions of this product

Catalog No. AAA1458503


Explore available promotions

Description

Description

2'-Hydroxy-4',6'-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-4′,6′-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2'-Hydroxy-4',6'-dimethoxyacetophenone
80°C to 83°C
C10H12O4
1 g
14,10067
Soluble in hot methanol.
COC1=CC(O)=C(C(C)=O)C(OC)=C1
196.20
CHEBI:10070
98%
90-24-4
185°C (20 mmHg)
MFCD00017243
1285013
xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
FBUBVLUPUDBFME-UHFFFAOYSA-N
1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
66654
196.2

Chemical Identifiers

90-24-4
196.20
FBUBVLUPUDBFME-UHFFFAOYSA-N
66654
1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
C10H12O4
MFCD00017243
xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
CHEBI:10070
COC1=CC(O)=C(C(C)=O)C(OC)=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-978-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only