missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

$103.04 - $337.73

Chemical Identifiers

CAS 90-24-4
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00017243
InChI Key FBUBVLUPUDBFME-UHFFFAOYSA-N
Synonym xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
PubChem CID 66654
ChEBI CHEBI:10070
IUPAC Name 1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
SMILES COC1=CC(O)=C(C(C)=O)C(OC)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1458503
View Documents
Thermo Scientific Chemicals
A1458503
1 g
Each for $103.04
 
AAA1458506
View Documents
Thermo Scientific Chemicals
A1458506
5 g
Each for $337.73
 
Description

Description

2'-Hydroxy-4',6'-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-4′,6′-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

90-24-4
196.20
FBUBVLUPUDBFME-UHFFFAOYSA-N
66654
1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
C10H12O4
MFCD00017243
xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
CHEBI:10070
COC1=CC(O)=C(C(C)=O)C(OC)=C1

Specifications

90-24-4
185°C (20 mmHg)
MFCD00017243
1285013
xanthoxylin, 2'-hydroxy-4',6'-dimethoxyacetophenone, brevifolin, xanthoxyline, 2-hydroxy-4,6-dimethoxyacetophenone, 1-2-hydroxy-4,6-dimethoxyphenyl ethanone, brevifolin zanthoxylum, phloracetophenone dimethyl ether, brevifolin van, 1-2-hydroxy-4,6-dimethoxyphenyl ethan-1-one
FBUBVLUPUDBFME-UHFFFAOYSA-N
1-(2-hydroxy-4,6-dimethoxyphenyl)ethanone
66654
196.2
2'-Hydroxy-4',6'-dimethoxyacetophenone
80°C to 83°C
C10H12O4
1 g
14,10067
Soluble in hot methanol.
COC1=CC(O)=C(C(C)=O)C(OC)=C1
196.20
CHEBI:10070
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-978-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only