missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxy-4-methoxybenzophenone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 121361000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Hydroxy-4-methoxybenzophenone | |
131-57-7 | |
100.0 | |
White to Yellow | |
>100°C | |
97.5% min. (GC) | |
C14H12O3 | |
MFCD00008387 | |
08,312 | |
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex | |
DXGLGDHPHMLXJC-UHFFFAOYSA-N | |
(2-hydroxy-4-methoxyphenyl)-phenylmethanone | |
4632 | |
228.25 | |
Crystalline Powder |
98% | |
97.5 | |
>62.5°C | |
150°C to 160°C (5.0 mmHg) | |
Authentic | |
Glass bottle | |
HOC6H3(OCH3)COC6H5 | |
100 g | |
15,7053 | |
Solubility in water: .. Other solubilities: readily soluble in most organic solvents | |
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O | |
228.25 | |
CHEBI:34283 | |
98% |
Chemical Identifiers
131-57-7 | |
228.25 | |
DXGLGDHPHMLXJC-UHFFFAOYSA-N | |
4632 | |
(2-hydroxy-4-methoxyphenyl)-phenylmethanone |
C14H12O3 | |
MFCD00008387 | |
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex | |
CHEBI:34283 | |
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O |
Safety and Handling
EINECSNumber : 205-031-5
RTECSNumber : DJ1575000
TSCA : TSCA
RUO – Research Use Only