missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-4-methoxybenzophenone, 98%, Thermo Scientific Chemicals

$51.48 - $106.13

Chemical Identifiers

CAS 131-57-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00008387
InChI Key DXGLGDHPHMLXJC-UHFFFAOYSA-N
Synonym oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
PubChem CID 4632
ChEBI CHEBI:34283
IUPAC Name (2-hydroxy-4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC121360050
SDS View Documents
Thermo Scientific Chemicals
121360050
5 g Glass bottle
Each for $51.48
 
AC121361000
SDS View Documents
Thermo Scientific Chemicals
121361000
100 g Glass bottle
Each for $106.13
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

131-57-7
228.25
DXGLGDHPHMLXJC-UHFFFAOYSA-N
4632
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
C14H12O3
MFCD00008387
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
CHEBI:34283
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

131-57-7
100.0
White to Yellow
>100°C
97.5% min. (GC)
C14H12O3
MFCD00008387
08,312
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
DXGLGDHPHMLXJC-UHFFFAOYSA-N
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
4632
228.25
Crystalline Powder
97.5
>62.5°C
150°C to 160°C (5.0 mmHg)
Authentic
Glass bottle
HOC6H3(OCH3)COC6H5
5 g
15,7053
Solubility in water: .. Other solubilities: readily soluble in most organic solvents
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
228.25
CHEBI:34283
98%
2-Hydroxy-4-methoxybenzophenone, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-031-5

RTECSNumber : DJ1575000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only