missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxy-5-nitrobenzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America H154925G
Specifications
2-Hydroxy-5-nitrobenzyl Alcohol | |
125°C | |
C7H7NO4 | |
25 g | |
JEIYIXDNZKATLC-UHFFFAOYSA-N | |
2-(hydroxymethyl)-4-nitrophenol | |
2763823 | |
≥98.0% (GC) |
39224-61-8 | |
Yellow | |
MFCD01321361 | |
5-Nitrosalicyl Alcohol | |
OCC1=CC(=CC=C1O)[N+]([O-])=O | |
169.14 | |
169.14 | |
Crystalline Powder |
Chemical Identifiers
39224-61-8 | |
169.14 | |
JEIYIXDNZKATLC-UHFFFAOYSA-N | |
2763823 | |
OCC1=CC(=CC=C1O)[N+]([O-])=O |
C7H7NO4 | |
MFCD01321361 | |
5-Nitrosalicyl Alcohol | |
2-(hydroxymethyl)-4-nitrophenol |
Safety and Handling
TSCA : No