missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-5-nitrobenzyl Alcohol 98.0+%, TCI America™
$136.08 - $473.48
Chemical Identifiers
| CAS | 39224-61-8 |
|---|---|
| Molecular Formula | C7H7NO4 |
| Molecular Weight (g/mol) | 169.14 |
| MDL Number | MFCD01321361 |
| InChI Key | JEIYIXDNZKATLC-UHFFFAOYSA-N |
| Synonym | 5-Nitrosalicyl Alcohol |
| PubChem CID | 2763823 |
| IUPAC Name | 2-(hydroxymethyl)-4-nitrophenol |
| SMILES | OCC1=CC(=CC=C1O)[N+]([O-])=O |
Chemical Identifiers
| 39224-61-8 | |
| 169.14 | |
| JEIYIXDNZKATLC-UHFFFAOYSA-N | |
| 2763823 | |
| OCC1=CC(=CC=C1O)[N+]([O-])=O |
| C7H7NO4 | |
| MFCD01321361 | |
| 5-Nitrosalicyl Alcohol | |
| 2-(hydroxymethyl)-4-nitrophenol |
Specifications
| 39224-61-8 | |
| Yellow | |
| MFCD01321361 | |
| 5-Nitrosalicyl Alcohol | |
| OCC1=CC(=CC=C1O)[N+]([O-])=O | |
| 169.14 | |
| 169.14 | |
| Crystalline Powder |
| 125°C | |
| C7H7NO4 | |
| 5 g | |
| JEIYIXDNZKATLC-UHFFFAOYSA-N | |
| 2-(hydroxymethyl)-4-nitrophenol | |
| 2763823 | |
| ≥98.0% (GC) | |
| 2-Hydroxy-5-nitrobenzyl Alcohol |
Safety and Handling
TSCA : No