Learn More
2-Hydroxy-6-methoxybenzaldehyde, 98+%, Thermo Scientific Chemicals
$134.10 - $484.05
Chemical Identifiers
CAS | 700-44-7 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
MDL Number | MFCD00151830 |
InChI Key | DZJPDDVDKXHRLF-UHFFFAOYSA-N |
PubChem CID | 69692 |
IUPAC Name | 2-hydroxy-6-methoxybenzaldehyde |
SMILES | COC1=CC=CC(=C1C=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1314503
|
Thermo Scientific Chemicals
L1314503 |
1 g |
N/A
|
|
|||||
AAL1314506
|
Thermo Scientific Chemicals
L1314506 |
5 g |
N/A
|
|
|||||
Description
2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
700-44-7 | |
152.149 | |
DZJPDDVDKXHRLF-UHFFFAOYSA-N | |
2-hydroxy-6-methoxybenzaldehyde |
C8H8O3 | |
MFCD00151830 | |
69692 | |
COC1=CC=CC(=C1C=O)O |
Specifications
700-44-7 | |
C8H8O3 | |
1 g | |
Air Sensitive | |
Slightly soluble in water. | |
COC1=CC=CC(=C1C=O)O | |
152.149 | |
152.15 | |
2-Hydroxy-6-methoxybenzaldehyde |
68°C to 74°C | |
MFCD00151830 | |
1101524 | |
6-methoxysalicylaldehyde, 2-hydroxy-6-methoxy-benzaldehyde, 2-hydroxy-6-methoxybenzal dehyde, 2-hydroxy-6-methoxy benzaldehyde, benzaldehyde, 2-hydroxy-6-methoxy, pubchem8210, 2-methoxy-6-hydroxybenzaldehyde, benzaldehyde,2-hydroxy-6-methoxy, 2-hydroxy-6-methyloxy benzaldehyde | |
DZJPDDVDKXHRLF-UHFFFAOYSA-N | |
2-hydroxy-6-methoxybenzaldehyde | |
69692 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-844-6
TSCA : No
RUO – Research Use Only