missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1393706

 View more versions of this product

Catalog No. AAL1393706


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2'-Hydroxychalcone
89°C to 91°C
MFCD00016441
976324
AETKQQBRKSELEL-ZHACJKMWSA-N
(E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
638276
224.26
1214-47-7
C15H12O2
5 g
2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O
224.259
CHEBI:27916
≥98%

Chemical Identifiers

1214-47-7
224.259
AETKQQBRKSELEL-ZHACJKMWSA-N
638276
(E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
C15H12O2
MFCD00016441
2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
CHEBI:27916
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-928-0

RTECSNumber : UD5579400

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only