missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals

$58.37 - $121.78

Chemical Identifiers

CAS 1214-47-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00016441
InChI Key AETKQQBRKSELEL-ZHACJKMWSA-N
Synonym 2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
PubChem CID 638276
ChEBI CHEBI:27916
IUPAC Name (E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1393706
View Documents
Thermo Scientific Chemicals
L1393706
5 g
Each for $58.37
 
AAL1393714
View Documents
Thermo Scientific Chemicals
L1393714
25 g
Each for $121.78
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1214-47-7
224.259
AETKQQBRKSELEL-ZHACJKMWSA-N
638276
(E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
C15H12O2
MFCD00016441
2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
CHEBI:27916
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O

Specifications

1214-47-7
C15H12O2
5 g
2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O
224.259
CHEBI:27916
≥98%
89°C to 91°C
MFCD00016441
976324
AETKQQBRKSELEL-ZHACJKMWSA-N
(E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
638276
224.26
2'-Hydroxychalcone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-928-0

RTECSNumber : UD5579400

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only