missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-N-methylbenzylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6333903

 View more versions of this product

Catalog No. AAH6333903


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Methoxy-N-methylbenzylamine
Colorless
234°C to 235°C
C9H13NO
MFCD00025755
UN2735
JCCQJCOMFAJJCQ-UHFFFAOYSA-N
1-(2-methoxyphenyl)-N-methylmethanamine
522302
97%
6851-80-5
1.015 g/mL
106°C (223°F)
1.5235
1 g
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, ksc495a5l
CNCC1=CC=CC=C1OC
151.209
151.21
Liquid

Chemical Identifiers

6851-80-5
151.209
JCCQJCOMFAJJCQ-UHFFFAOYSA-N
522302
CNCC1=CC=CC=C1OC
C9H13NO
MFCD00025755
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, ksc495a5l
1-(2-methoxyphenyl)-N-methylmethanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions