Learn More
2-Methoxy-N-methylbenzylamine, 97%, Thermo Scientific™
$85.92 - $85.92
Chemical Identifiers
CAS | 6851-80-5 |
---|---|
Molecular Formula | C9H13NO |
Molecular Weight (g/mol) | 151.209 |
MDL Number | MFCD00025755 |
InChI Key | JCCQJCOMFAJJCQ-UHFFFAOYSA-N |
Synonym | 2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, ksc495a5l |
PubChem CID | 522302 |
IUPAC Name | 1-(2-methoxyphenyl)-N-methylmethanamine |
SMILES | CNCC1=CC=CC=C1OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6333903
|
Thermo Scientific Chemicals
H6333903 |
1 g |
Each for $85.92
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6851-80-5 | |
151.209 | |
JCCQJCOMFAJJCQ-UHFFFAOYSA-N | |
522302 | |
CNCC1=CC=CC=C1OC |
C9H13NO | |
MFCD00025755 | |
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, ksc495a5l | |
1-(2-methoxyphenyl)-N-methylmethanamine |
Specifications
6851-80-5 | |
1.015 g/mL | |
106°C (223°F) | |
1.5235 | |
1 g | |
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, ksc495a5l | |
CNCC1=CC=CC=C1OC | |
151.209 | |
151.21 | |
Liquid |
Colorless | |
234°C to 235°C | |
C9H13NO | |
MFCD00025755 | |
UN2735 | |
JCCQJCOMFAJJCQ-UHFFFAOYSA-N | |
1-(2-methoxyphenyl)-N-methylmethanamine | |
522302 | |
97% | |
2-Methoxy-N-methylbenzylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures