missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxypropene, 95%, stab. with ca 0.5% potassium carbonate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2408318

 View more versions of this product

Catalog No. AAB2408318


Explore available promotions

Description

Description

It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container.
Specifications

Specifications

2-Methoxypropene
116-11-0
35°C to 36°C
Ether-like
1.3835
50 g
1734635
2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
YOWQWFMSQCOSBA-UHFFFAOYSA-N
2-methoxyprop-1-ene
8300
95%
stabilized with ≈0.5% potassium carbonate
0.758
−29°C (−20°F)
C4H8O
MFCD00014929
UN1993
Light sensitive
Insoluble in water.
CC(=C)OC
72.107
72.11

Chemical Identifiers

116-11-0
72.107
YOWQWFMSQCOSBA-UHFFFAOYSA-N
8300
CC(=C)OC
C4H8O
MFCD00014929
2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
2-methoxyprop-1-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H301
Extremely flammable liquid and vapour.
Toxic if swallowed.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c

H224-H302

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 204-125-3

RTECSNumber : UD0800000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only