Learn More
2-Methoxypropene, 95%, stab. with ca 0.5% potassium carbonate, Thermo Scientific Chemicals
$86.58 - $279.41
Chemical Identifiers
CAS | 116-11-0 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.107 |
MDL Number | MFCD00014929 |
InChI Key | YOWQWFMSQCOSBA-UHFFFAOYSA-N |
Synonym | 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b |
PubChem CID | 8300 |
IUPAC Name | 2-methoxyprop-1-ene |
SMILES | CC(=C)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2408318
|
Thermo Scientific Chemicals
B2408318 |
50 g |
Each for $86.58
|
|
|||||
AAB2408330
|
Thermo Scientific Chemicals
B2408330 |
250 g |
Each for $279.41
|
|
|||||
Description
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used for Synthesis of heterocycle compounds and an important intermediate of medicine.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in a cool, dry place in tightly closed container.
Chemical Identifiers
116-11-0 | |
72.107 | |
YOWQWFMSQCOSBA-UHFFFAOYSA-N | |
8300 | |
CC(=C)OC |
C4H8O | |
MFCD00014929 | |
2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b | |
2-methoxyprop-1-ene |
Specifications
116-11-0 | |
35°C to 36°C | |
Ether-like | |
1.3835 | |
50 g | |
1734635 | |
2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b | |
YOWQWFMSQCOSBA-UHFFFAOYSA-N | |
2-methoxyprop-1-ene | |
8300 | |
95% |
0.758 | |
−29°C (−20°F) | |
C4H8O | |
MFCD00014929 | |
UN1993 | |
Light sensitive | |
Insoluble in water. | |
CC(=C)OC | |
72.107 | |
72.11 | |
2-Methoxypropene, stabilized with ≈0.5% potassium carbonate |
Safety and Handling
GHS H Statement
H224-H301
Extremely flammable liquid and vapour.
Toxic if swallowed.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H224-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 204-125-3
RTECSNumber : UD0800000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only