missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1,4-naphthoquinone, 98%, Thermo Scientific Chemicals

$77.72 - $303.66

Chemical Identifiers

CAS 58-27-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD00001681
InChI Key MJVAVZPDRWSRRC-UHFFFAOYSA-N
Synonym menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
PubChem CID 4055
ChEBI CHEBI:28869
IUPAC Name 2-methylnaphthalene-1,4-dione
SMILES CC1=CC(=O)C2=CC=CC=C2C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127180250
SDS View Documents
Thermo Scientific Chemicals
127180250
25 g Glass bottle
Each for $77.72
 
AC127181000
SDS View Documents
Thermo Scientific Chemicals
127181000
100 g Glass bottle
Each for $303.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58-27-5
172.18
MJVAVZPDRWSRRC-UHFFFAOYSA-N
4055
2-methylnaphthalene-1,4-dione
C11H8O2
MFCD00001681
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
CHEBI:28869
CC1=CC(=O)C2=CC=CC=C2C1=O

Specifications

58-27-5
100.0
Green-Yellow to Yellow
97.5% min. (HPLC)
C11H8O2
25 g
15, 5899
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol (1g/60 mL),benzene (1g/10 mL,soluble in vegetable oils (1g/50 mL),moderately soluble in chloroform and carbon tetrac
CC1=CC(=O)C2=CC=CC=C2C1=O
172.18
CHEBI:28869
98%
2-Methyl-1, 4-naphthoquinone, 98%
97.5
105.0°C to 108.0°C
Authentic
Glass bottle
MFCD00001681
07, II, 656
menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
MJVAVZPDRWSRRC-UHFFFAOYSA-N
2-methylnaphthalene-1,4-dione
4055
172.18
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 200-372-6

RTECSNumber : QL9100000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only