Learn More
2-Methyl-1-phenyl-1-propanol, 98%, Thermo Scientific Chemicals
$180.58 - $674.86
Chemical Identifiers
CAS | 611-69-8 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.221 |
MDL Number | MFCD00065000 |
InChI Key | GMDYDZMQHRTHJA-UHFFFAOYSA-N |
Synonym | 2-methyl-1-phenyl-1-propanol, .alpha.-isopropylbenzyl alcohol, isopropyl benzyl alcohol, 2-methyl-1-phenylpropanol, benzenemethanol, .alpha.-1-methylethyl, 1-phenyl-2-methylpropyl alcohol, 1r-2-methyl-1-phenyl-propan-1-ol, phenylisopropylcarbinol, isopropylphenylcarbinol, acmc-20anr9 |
PubChem CID | 95626 |
IUPAC Name | 2-methyl-1-phenylpropan-1-ol |
SMILES | CC(C)C(C1=CC=CC=C1)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2072906
|
Thermo Scientific Chemicals
B2072906 |
5 g |
Each for $180.58
|
|
|||||
AAB2072914
|
Thermo Scientific Chemicals
B2072914 |
25 g |
Each for $674.86
|
|
|||||
Description
It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker's yeast mediated asymmetric reduction of cinnamaldehyde derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker′s yeast mediated asymmetric reduction of cinnamaldehyde derivatives.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
611-69-8 | |
150.221 | |
GMDYDZMQHRTHJA-UHFFFAOYSA-N | |
95626 | |
CC(C)C(C1=CC=CC=C1)O |
C10H14O | |
MFCD00065000 | |
2-methyl-1-phenyl-1-propanol, .alpha.-isopropylbenzyl alcohol, isopropyl benzyl alcohol, 2-methyl-1-phenylpropanol, benzenemethanol, .alpha.-1-methylethyl, 1-phenyl-2-methylpropyl alcohol, 1r-2-methyl-1-phenyl-propan-1-ol, phenylisopropylcarbinol, isopropylphenylcarbinol, acmc-20anr9 | |
2-methyl-1-phenylpropan-1-ol |
Specifications
611-69-8 | |
124°C to 125°C (15 mmHg) | |
C10H14O | |
MFCD00065000 | |
2-methyl-1-phenyl-1-propanol, .alpha.-isopropylbenzyl alcohol, isopropyl benzyl alcohol, 2-methyl-1-phenylpropanol, benzenemethanol, .alpha.-1-methylethyl, 1-phenyl-2-methylpropyl alcohol, 1r-2-methyl-1-phenyl-propan-1-ol, phenylisopropylcarbinol, isopropylphenylcarbinol, acmc-20anr9 | |
GMDYDZMQHRTHJA-UHFFFAOYSA-N | |
2-methyl-1-phenylpropan-1-ol | |
95626 | |
98% |
0.964 | |
86°C (187°F) | |
1.513 | |
5 g | |
Not miscible or difficult to mix in water. | |
CC(C)C(C1=CC=CC=C1)O | |
150.221 | |
150.22 | |
2-Methyl-1-phenyl-1-propanol |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 210-274-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only