missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol, 99+%, Extra Pure

2-Methyl-1-propanol, 99+%, C4H10O, CAS Number-78-83-1, 1-hydroxymethylpropane, 1-propanol, 2-methyl, 2-methyl-1-propanol, 2-methylpropyl alcohol, i-butyl alcohol, isobutanol, iso-butyl alcohol, isobutyl alcohol, isobutylalkohol, isopropylcarbinol, 2.5L, 99% min. (GC), 01, 373, 108 deg.C, 100.0, 99.0 | CAS: 78-83-1 | C4H10O | 74.12 g/mol

Supplier:  Thermo Scientific Chemicals 158280010

 View more versions of this product

Catalog No. AC158280010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-1-propanol
99.0
-108°C
108°C
Authentic
Glass bottle
1.3945 to 1.3965
MFCD00004740
01, 373
0.8
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
2-methylpropan-1-ol
6560
CHEBI:46645
11.7mbar at 20°C
Extra Pure
78-83-1
100.0
0.8000g/mL
28°C
99% min. (GC)
C4H10O
(CH3)2CHCH2OH
1 L
0.005% max.
15, 5176
Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone
CC(C)CO
74.12
4 Pas (20°C)
74.12
99+%
Liquid

Chemical Identifiers

78-83-1
74.12
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
6560
2-methylpropan-1-ol
C4H10O
MFCD00004740
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
CHEBI:46645
CC(C)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at

GHS Signal Word: Danger

EINECSNumber : 201-148-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only