Learn More
2-Methyl-1-propanol, 99+%, Extra Pure, Thermo Scientific Chemicals
2-Methyl-1-propanol, 99+%, C4H10O, CAS Number-78-83-1, 1-hydroxymethylpropane, 1-propanol, 2-methyl, 2-methyl-1-propanol, 2-methylpropyl alcohol, i-butyl alcohol, isobutanol, iso-butyl alcohol, isobutyl alcohol, isobutylalkohol, isopropylcarbinol, 2.5L, 99% min. (GC), 01, 373, 108 deg.C, 100.0, 99.0
$107.72 - $193.09
Chemical Identifiers
CAS | 78-83-1 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00004740 |
InChI Key | ZXEKIIBDNHEJCQ-UHFFFAOYSA-N |
Synonym | 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol |
PubChem CID | 6560 |
ChEBI | CHEBI:46645 |
IUPAC Name | 2-methylpropan-1-ol |
SMILES | CC(C)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC158280010
|
Thermo Scientific Chemicals
158280010 |
1 L | Glass bottle |
Each for $107.72
|
|
||||
AC158280025
|
Thermo Scientific Chemicals
158280025 |
2.5 L | Glass bottle |
Each for $193.09
Case of 4 Each for $0.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-83-1 | |
74.12 | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
6560 | |
2-methylpropan-1-ol |
C4H10O | |
MFCD00004740 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
CHEBI:46645 | |
CC(C)CO |
Specifications
78-83-1 | |
100.0 | |
0.8000g/mL | |
28°C | |
99% min. (GC) | |
C4H10O | |
(CH3)2CHCH2OH | |
1 L | |
0.005% max. | |
15, 5176 | |
Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone | |
CC(C)CO | |
74.12 | |
4 Pas (20°C) | |
74.12 | |
99+% | |
Liquid |
99.0 | |
-108°C | |
108°C | |
Authentic | |
Glass bottle | |
1.3945 to 1.3965 | |
MFCD00004740 | |
01, 373 | |
0.8 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
2-methylpropan-1-ol | |
6560 | |
CHEBI:46645 | |
11.7mbar at 20°C | |
Extra Pure | |
2-Methyl-1-propanol |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
GHS Signal Word: Danger
EINECSNumber : 201-148-
RUO – Research Use Only